Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193760.2 | NP_001180689.2 | 786 | 142 | ||
XM_015125328.2 | XP_014980814.2 | 1180 | 142 | |||
GenBank mRNA | JU493918.1 | - | 782 | 142 |

More Information
Set Membership: |
![]() ![]() |
