Assay Details
| Gene Symbol: TTI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193802.3 | NP_001180731.2 | 1684 | 112 | ||
| XM_015145053.2 | XP_015000539.1 | 1667 | 112 | |||
| GenBank mRNA | JU336483.1 | - | 1324 | 112 | ||
| JU472787.1 | - | 1423 | 112 | |||
| JV044754.1 | - | 1447 | 112 | |||
More Information
Set Membership: |
|

TTI2
Genomic Map