Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265820.1 | NP_001252749.1 | 307 | 103 | ||
| XM_015152975.2 | XP_015008461.1 | 406 | 103 | |||
| GenBank mRNA | JV667482.1 | - | 307 | 103 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF34
Genomic Map