Assay Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194197.2 | NP_001181126.1 | 289 | 75 | ||
| XM_001101661.4 | XP_001101661.1 | 621 | 75 | |||
| XM_015153074.2 | XP_015008560.1 | 714 | 75 | |||
| GenBank mRNA | JU321383.1 | - | 321 | 75 | ||
| JV048306.1 | - | 310 | 75 | |||
More Information
Set Membership: |
|

CCDC92
Genomic Map