Assay Details
Gene Symbol: ZNF436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257465.1 | NP_001244394.1 | 387 | 98 | ||
XM_028843650.1 | XP_028699483.1 | 2512 | 98 | |||
GenBank mRNA | JU336521.1 | - | 387 | 98 | ||
JU477330.1 | - | 417 | 98 | |||
JV048323.1 | - | 595 | 98 | |||
JV048324.1 | - | 406 | 98 | |||
JV662371.1 | - | 586 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257465.1, XM_028843650.1 transcripts.
Set Membership: |
![]() ![]() |
