Assay Details
Gene Symbol: LIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116341.4 | XP_001116341.1 | 501 | 60 | ||
XM_001116349.4 | XP_001116349.1 | 627 | 60 | |||
XM_028839119.1 | XP_028694952.1 | 600 | 60 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
