Assay Details
| Gene Symbol: MAP2K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122493.2 | XP_014977979.1 | 1360 | 104 | ||
| XM_015122494.2 | XP_014977980.1 | 1213 | 104 | |||
| GenBank mRNA | JU322288.1 | - | 1093 | 104 | ||
| JU470476.1 | - | 1190 | 104 | |||
| JU470477.1 | - | 1190 | 104 | |||
| JV045089.1 | - | 1107 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAP2K2
Genomic Map