Assay Details
| Gene Symbol: ZSCAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135802.2 | XP_014991288.2 | 3475 | 132 | ||
| XM_028847753.1 | XP_028703586.1 | 2488 | 132 | |||
| GenBank mRNA | JU347109.1 | - | 656 | 132 | ||
More Information
Set Membership: |
|

ZSCAN31
Genomic Map