Assay Details
| Gene Symbol: SHARPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146258.2 | XP_015001744.1 | 1290 | 114 | ||
| XM_015146259.2 | XP_015001745.1 | 1290 | 114 | |||
| XM_015146260.2 | XP_015001746.1 | 1287 | 114 | |||
| XM_015146261.2 | XP_015001747.1 | 1290 | 114 | |||
| XM_015146263.2 | XP_015001749.1 | 1290 | 114 | |||
| XM_028853111.1 | XP_028708944.1 | 1290 | 114 | |||
| XM_028853112.1 | XP_028708945.1 | 1185 | 114 | |||
| GenBank mRNA | JV595040.1 | - | 1211 | 114 | ||
| JV636394.1 | - | 1183 | 114 | |||
| JV724421.1 | - | 745 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SHARPIN
Genomic Map