Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194157.2 | NP_001181086.1 | 538 | 154 | ||
| XM_001100385.4 | XP_001100385.1 | 944 | 154 | |||
| XM_015122452.2 | XP_014977938.1 | 2496 | 154 | |||
| XM_015122453.2 | XP_014977939.1 | 599 | 154 | |||
| GenBank mRNA | JU322507.1 | - | 485 | 154 | ||
| JU471205.1 | - | 528 | 154 | |||
| JV602054.1 | - | 531 | 154 | |||
| JV704522.1 | - | 510 | 154 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DOHH
Genomic Map