Assay Details
Gene Symbol: MRPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265691.1 | NP_001252620.1 | 629 | 88 | ||
XM_028844350.1 | XP_028700183.1 | 1240 | 88 | |||
GenBank mRNA | JU325541.1 | - | 579 | 88 | ||
JV600316.1 | - | 629 | 88 | |||
JV669320.1 | - | 646 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
