Assay Details
Gene Symbol: LOC713879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105834.3 | XP_001105834.1 | 1211 | 155 | ||
XM_015123623.2 | XP_014979109.1 | 774 | 155 | |||
XM_015123624.2 | XP_014979110.1 | 387 | 155 | |||
XM_015123625.2 | XP_014979111.1 | 436 | 155 | |||
XM_028839052.1 | XP_028694885.1 | 464 | 155 | |||
GenBank mRNA | JV052696.1 | - | 423 | 155 | ||
JV643217.1 | - | 423 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() |
