Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261475.1 | NP_001248404.1 | 496 | 88 | ||
| XM_015150645.2 | XP_015006131.1 | 1742 | 88 | |||
| XM_015150663.2 | XP_015006149.1 | 1661 | 88 | |||
| XM_015150685.2 | XP_015006171.1 | 1436 | 88 | |||
| XM_028843230.1 | XP_028699063.1 | 224 | 88 | |||
| GenBank mRNA | JU323064.1 | - | 496 | 88 | ||
More Information
Set Membership: |
|

CIART
Genomic Map