Assay Details
Gene Symbol: SEPTIN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113104.2 | XP_014968590.1 | 469 | 114 | ||
XM_015113113.2 | XP_014968599.1 | 423 | 114 | |||
XM_028832293.1 | XP_028688126.1 | 469 | 114 | |||
GenBank mRNA | JU321498.1 | - | 173 | 114 | ||
JV638942.1 | - | 76 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015113104.2, XM_015113113.2, XM_028832293.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
