Assay Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129222.2 | XP_014984708.1 | 964 | 132 | ||
XM_015129230.2 | XP_014984716.2 | 639 | 132 | |||
XM_015129235.2 | XP_014984721.1 | 899 | 132 | |||
XM_015129242.2 | XP_014984728.1 | 654 | 132 | |||
XM_028844705.1 | XP_028700538.1 | 1095 | 132 | |||
XM_028844706.1 | XP_028700539.1 | 1095 | 132 | |||
XM_028844707.1 | XP_028700540.1 | 767 | 132 | |||
GenBank mRNA | JU323036.1 | - | 957 | 132 | ||
JU474618.1 | - | 932 | 132 | |||
JV045461.1 | - | 872 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
