Assay Details
| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826225.1 | XP_028682058.1 | 151 | 97 | ||
| XM_028826227.1 | XP_028682060.1 | 141 | 97 | |||
| GenBank mRNA | JU323889.1 | - | 30 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826225.1, XM_028826227.1 transcripts.
Set Membership: |
|

ZNF25
Genomic Map