Assay Details
Gene Symbol: ZNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096234.4 | XP_001096234.3 | 785 | 79 | ||
XM_028826224.1 | XP_028682057.1 | 538 | 79 | |||
XM_028826225.1 | XP_028682058.1 | 370 | 79 | |||
XM_028826226.1 | XP_028682059.1 | 359 | 79 | |||
XM_028826227.1 | XP_028682060.1 | 383 | 79 | |||
GenBank mRNA | JU323889.1 | - | 249 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826227.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
