Assay Details
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115343.2 | XP_014970829.2 | 558 | 93 | ||
XM_028833876.1 | XP_028689709.1 | 232 | 93 | |||
GenBank mRNA | JU482470.1 | - | 77 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
