Assay Details
| Gene Symbol: ZNF627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107785.4 | XP_001107785.2 | 396 | 139 | ||
| GenBank mRNA | JU330075.1 | - | 177 | 139 | ||
| JV644625.1 | - | 373 | 139 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF627
Genomic Map