Assay Details
| Gene Symbol: LTC4S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101985.4 | XP_001101985.2 | 345 | 72 | ||
| XM_028850008.1 | XP_028705841.1 | 345 | 72 | |||
| GenBank mRNA | JU334499.1 | - | 240 | 72 | ||
| JV651474.1 | - | 251 | 72 | |||
More Information
Set Membership: |
|

LTC4S
Genomic Map