Assay Details
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097066.4 | XP_001097066.1 | 633 | 114 | ||
XM_028850289.1 | XP_028706122.1 | 624 | 114 | |||
XM_028850290.1 | XP_028706123.1 | 670 | 114 | |||
XM_028850292.1 | XP_028706125.1 | 1876 | 114 | |||
XM_028850296.1 | XP_028706129.1 | 674 | 114 | |||
XM_028850297.1 | XP_028706130.1 | 1763 | 114 | |||
XM_028850300.1 | XP_028706133.1 | 808 | 114 | |||
GenBank mRNA | JU350595.1 | - | 565 | 114 | ||
JV726922.1 | - | 572 | 114 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028850289.1, XM_028850290.1, XM_028850292.1, XM_028850296.1, XM_028850297.1, XM_028850300.1 transcripts.
This assay is in the 5'UTR region of the XM_028850289.1, XM_028850290.1, XM_028850292.1, XM_028850296.1, XM_028850297.1, XM_028850300.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
