Assay Details
| Gene Symbol: LOC716404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836627.1 | XP_028692460.1 | 2691 | 115 | ||
| XM_028836628.1 | XP_028692461.1 | 2208 | 115 | |||
More Information
Set Membership: |
|

LOC716404
Genomic Map