Assay Details
Gene Symbol: CCDC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028834552.1 | XP_028690385.1 | 885 | 83 | ||
XM_028834555.1 | XP_028690388.1 | 882 | 83 | |||
XM_028834557.1 | XP_028690390.1 | 885 | 83 | |||
XM_028834561.1 | XP_028690394.1 | 882 | 83 | |||
XM_028834567.1 | XP_028690400.1 | 846 | 83 | |||
XM_028834572.1 | XP_028690405.1 | 843 | 83 | |||
XM_028834574.1 | XP_028690407.1 | 846 | 83 | |||
XM_028834579.1 | XP_028690412.1 | 843 | 83 | |||
XM_028834588.1 | XP_028690421.1 | 627 | 83 | |||
XM_028834590.1 | XP_028690423.1 | 551 | 83 | |||
XM_028834593.1 | XP_028690426.1 | 554 | 83 | |||
XM_028834598.1 | XP_028690431.1 | 512 | 83 | |||
XM_028834602.1 | XP_028690435.1 | 512 | 83 | |||
XM_028834603.1 | XP_028690436.1 | 619 | 83 | |||
XM_028834609.1 | XP_028690442.1 | 357 | 83 | |||
GenBank mRNA | JU474422.1 | - | 571 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
