Assay Details
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117775.2 | XP_014973261.2 | 2448 | 95 | ||
XM_015117777.2 | XP_014973263.2 | 3894 | 95 | |||
XM_015117778.2 | XP_014973264.2 | 2448 | 95 | |||
XM_015117779.2 | XP_014973265.2 | 2448 | 95 | |||
XM_028835185.1 | XP_028691018.1 | 2077 | 95 | |||
XM_028835186.1 | XP_028691019.1 | 1700 | 95 | |||
GenBank mRNA | JV053561.1 | - | 420 | 95 | ||
JV644096.1 | - | 306 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
