Assay Details
| Gene Symbol: BEND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257859.2 | NP_001244788.1 | 498 | 94 | ||
| XM_015136544.2 | XP_014992030.1 | 244 | 94 | |||
| XM_015136545.2 | XP_014992031.1 | 237 | 94 | |||
| XM_028846798.1 | XP_028702631.1 | 1404 | 94 | |||
| XM_028846799.1 | XP_028702632.1 | 238 | 94 | |||
| XM_028846800.1 | XP_028702633.1 | 702 | 94 | |||
| GenBank mRNA | JU333401.1 | - | 498 | 94 | ||
| JV608936.1 | - | 513 | 94 | |||
More Information
Set Membership: |
|

BEND6
Genomic Map