Assay Details
| Gene Symbol: AKNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086115.4 | XP_001086115.2 | 1449 | 122 | ||
| XM_001086220.4 | XP_001086220.2 | 1449 | 122 | |||
| XM_015146430.2 | XP_015001916.2 | 1449 | 122 | |||
| XM_015146447.2 | XP_015001933.2 | 1449 | 122 | |||
| XM_015146461.2 | XP_015001947.2 | 1449 | 122 | |||
| XM_028830849.1 | XP_028686682.1 | 1444 | 122 | |||
More Information
Set Membership: |
|

AKNAD1
Genomic Map