Assay Details
| Gene Symbol: GPR26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265705.1 | NP_001252634.1 | 817 | 153 | ||
| GenBank mRNA | JV600222.1 | - | 817 | 153 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR26| Gene Symbol: GPR26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265705.1 | NP_001252634.1 | 817 | 153 | ||
| GenBank mRNA | JV600222.1 | - | 817 | 153 | ||
Set Membership: |
|