Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099752.4 | XP_001099752.2 | 1101 | 104 | ||
XM_015140269.2 | XP_014995755.1 | 1101 | 104 | |||
GenBank mRNA | JU478157.1 | - | 747 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
