Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083815.4 | XP_001083815.1 | 622 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001083815.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
