Assay Details
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109787.4 | XP_001109787.2 | 579 | 110 | ||
XM_028827844.1 | XP_028683677.1 | 626 | 110 | |||
GenBank mRNA | JV648954.1 | - | 487 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
