Assay Details
| Gene Symbol: JAZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845920.1 | XP_028701753.1 | 785 | 83 | ||
| GenBank mRNA | JU333412.1 | - | 640 | 83 | ||
| JU474698.1 | - | 647 | 83 | |||
| JV608993.1 | - | 649 | 83 | |||
| JV645327.1 | - | 654 | 83 | |||
More Information
Set Membership: |
|

JAZF1
Genomic Map