Assay Details
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130375.2 | XP_014985861.2 | 5559 | 78 | ||
XM_015130376.2 | XP_014985862.2 | 3217 | 78 | |||
XM_028844377.1 | XP_028700210.1 | 3221 | 78 | |||
XM_028844378.1 | XP_028700211.1 | 3169 | 78 | |||
XM_028844379.1 | XP_028700212.1 | 3351 | 78 | |||
XM_028844380.1 | XP_028700213.1 | 3247 | 78 | |||
GenBank mRNA | JV707905.1 | - | 2955 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
