Assay Details
| Gene Symbol: MRPL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265996.2 | NP_001252925.1 | 224 | 88 | ||
| XM_015143274.2 | XP_014998760.2 | 251 | 88 | |||
| XM_015143278.2 | XP_014998764.2 | 372 | 88 | |||
| GenBank mRNA | JV489155.1 | - | 224 | 88 | ||
More Information
Set Membership: |
|

MRPL52
Genomic Map