Assay Details
Gene Symbol: SAPCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091379.4 | XP_001091379.4 | 760 | 73 | ||
XM_028834573.1 | XP_028690406.1 | 2196 | 73 | |||
GenBank mRNA | JU495809.1 | - | 82 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
