Assay Details
| Gene Symbol: LOC707884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090663.4 | XP_001090663.1 | 92 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001090663.4 transcripts.
Set Membership: |
|

LOC707884
Genomic Map