Assay Details
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848351.1 | XP_028704184.1 | 3068 | 82 | ||
XM_028848352.1 | XP_028704185.1 | 3049 | 82 | |||
XM_028848353.1 | XP_028704186.1 | 2992 | 82 | |||
XM_028848354.1 | XP_028704187.1 | 2983 | 82 | |||
XM_028848355.1 | XP_028704188.1 | 2968 | 82 | |||
XM_028848356.1 | XP_028704189.1 | 2959 | 82 | |||
XM_028848357.1 | XP_028704190.1 | 1288 | 82 | |||
GenBank mRNA | JU325974.1 | - | 1292 | 82 | ||
JU326050.1 | - | 1382 | 82 | |||
JU327732.1 | - | 1358 | 82 | |||
JV571476.1 | - | 1391 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
