Assay Details
| Gene Symbol: TMPRSS11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138610.2 | XP_014994096.1 | 373 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11B| Gene Symbol: TMPRSS11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138610.2 | XP_014994096.1 | 373 | 107 | ||
Set Membership: |
|