Assay Details
| Gene Symbol: CNIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113644.2 | XP_014969130.1 | 438 | 85 | ||
| XM_028833213.1 | XP_028689046.1 | 440 | 85 | |||
| GenBank mRNA | JU333800.1 | - | 206 | 85 | ||
| JU473574.1 | - | 198 | 85 | |||
| JV679109.1 | - | 288 | 85 | |||
| JV679110.1 | - | 284 | 85 | |||
More Information
Set Membership: |
|

CNIH2
Genomic Map