Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194053.2 | NP_001180982.2 | 192 | 159 | ||
| GenBank mRNA | JU342097.1 | - | 260 | 159 | ||
| JU478354.1 | - | 221 | 159 | |||
More Information
Set Membership: |
|

VMO1
Genomic Map