Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121526.2 | XP_014977012.2 | 749 | 91 | ||
| GenBank mRNA | JV059999.1 | - | 695 | 91 | ||
| JV635832.1 | - | 693 | 91 | |||
More Information
Set Membership: |
|

TMEM255B
Genomic Map