Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142997.2 | XP_014998483.1 | 316 | 127 | ||
XM_015142998.2 | XP_014998484.1 | 316 | 127 | |||
GenBank mRNA | JU323210.1 | - | 284 | 127 | ||
JV045082.1 | - | 200 | 127 | |||
JV580634.1 | - | 295 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
