Assay Details
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096810.4 | XP_001096810.2 | 648 | 69 | ||
XM_015147579.2 | XP_015003065.2 | 1053 | 69 | |||
XM_028826616.1 | XP_028682449.1 | 927 | 69 | |||
XM_028826617.1 | XP_028682450.1 | 978 | 69 | |||
XM_028826618.1 | XP_028682451.1 | 900 | 69 | |||
XM_028826619.1 | XP_028682452.1 | 874 | 69 | |||
GenBank mRNA | JU324011.1 | - | 350 | 69 | ||
JV645225.1 | - | 403 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
