Assay Details
| Gene Symbol: HACD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261129.1 | NP_001248058.1 | 171 | 73 | ||
| XM_028843389.1 | XP_028699222.1 | 197 | 73 | |||
| GenBank mRNA | JU327265.1 | - | 146 | 73 | ||
| JV044709.1 | - | 171 | 73 | |||
| JV639352.1 | - | 167 | 73 | |||

HACD2
Genomic Map