Assay Details
Gene Symbol: C19H19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194075.3 | NP_001181004.2 | 641 | 93 | ||
XM_015123960.2 | XP_014979446.2 | 1562 | 93 | |||
XM_028838387.1 | XP_028694220.1 | 982 | 93 | |||
XM_028838388.1 | XP_028694221.1 | 965 | 93 | |||
XM_028838389.1 | XP_028694222.1 | 680 | 93 | |||
GenBank mRNA | JV636107.1 | - | 761 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
