Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194823.2 | NP_001181752.1 | 273 | 82 | ||
| XM_001104593.4 | XP_001104593.1 | 405 | 82 | |||
| XM_001104745.4 | XP_001104745.1 | 469 | 82 | |||
| XM_001104890.4 | XP_001104890.1 | 555 | 82 | |||
| XM_015122915.2 | XP_014978401.1 | 504 | 82 | |||
| XM_015122917.2 | XP_014978403.1 | 440 | 82 | |||
| XM_015122918.2 | XP_014978404.1 | 357 | 82 | |||
| GenBank mRNA | JU321070.1 | - | 880 | 82 | ||
| JU334413.1 | - | 430 | 82 | |||
| JU471808.1 | - | 684 | 82 | |||
| JV044139.1 | - | 591 | 82 | |||
| JV044140.1 | - | 391 | 82 | |||
| JV452451.1 | - | 852 | 82 | |||
More Information
Set Membership: |
|

TMEM205
Genomic Map