Assay Details
| Gene Symbol: STIMATE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130713.2 | XP_014986199.1 | 700 | 112 | ||
| GenBank mRNA | JU327281.1 | - | 434 | 112 | ||
| JV044746.1 | - | 614 | 112 | |||
| JV636138.1 | - | 607 | 112 | |||
| JV714366.1 | - | 435 | 112 | |||
More Information
Set Membership: |
|

STIMATE
Genomic Map