Assay Details
| Gene Symbol: NDUFAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194523.1 | NP_001181452.1 | 936 | 98 | ||
| GenBank mRNA | JU323160.1 | - | 931 | 98 | ||
| JU470501.1 | - | 939 | 98 | |||
| JV046683.1 | - | 488 | 98 | |||
| JV540270.1 | - | 934 | 98 | |||
More Information
Set Membership: |
|

NDUFAF3
Genomic Map