Assay Details
Gene Symbol: ATPSCKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139734.2 | XP_014995220.2 | 776 | 162 | ||
GenBank mRNA | JU338908.1 | - | 492 | 162 | ||
JU473054.1 | - | 508 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() |
