Assay Details
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080768.1 | NP_001074237.1 | 105 | 56 | ||
GenBank mRNA | JU323214.1 | - | 139 | 56 | ||
JU473384.1 | - | 170 | 56 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
