Assay Details
| Gene Symbol: CHCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258084.1 | NP_001245013.1 | 137 | 106 | ||
| GenBank mRNA | JU474254.1 | - | 137 | 106 | ||
| JV046146.1 | - | 139 | 106 | |||
More Information
Set Membership: |
|

CHCHD1
Genomic Map